Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Powdered Agarose (Low EEO)
SCI_ED

Catalog No. S37746 Shop All Aldon Products
Change view
Click to view available options
Grade:
7 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S37746 7 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S37746 Supplier Innovating Science™ Supplier No. IS5205
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prepare your own agarose gels at any concentration with the addition of TBE running buffer.

  • Can be stored at room temperature for years

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) To prepare own agarose gels at any concentration with just addition of TBE running buffer
Includes Powdered Agarose (Low EEO)
Quantity 25 g
Dimensions (L x W x H) 1.5 x 1.5 x 3 in.
Product Type Powdered Agarose (Low EEO)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.