Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alconox™ Powdered Precision Cleaner
GREENER_CHOICE

Catalog No. 16000106 Shop All Alconox Products
Change view
Click to view available options
Volume (English):
4 lb.
25 lb.
50 x 0.5 oz.
50 lb.
100 lb.
300 lb.
Container Type:
Box
Carton
Drum
Packets
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English) Container Type
16-000-106 50 lb. Carton
16-000-107 100 lb. Drum
16-000-105 25 lb. Carton
16-000-104 4 lb. Box
16-000-108 300 lb. Drum
16-000-109 50 x 0.5 oz. Packets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 16-000-106 Supplier Alconox™ Supplier No. 1150
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Concentrated, anionic detergent for manual and ultrasonic cleaning

Alconox consists primarily of a homogeneous blend of sodium linear alkylaryl sulfonate, alcohol sulfate, phosphates, carbonates.

  • Biodegradable and readily disposable
  • Penetrating wetting power to save time
  • Replaces corrosive acids and hazardous solvents
  • Free rinsing gives reliable results without interfering residues
  • Use to pass cleaning validation tests for lab accreditation and plant inspection approval
  • Dilute 1:100 for typical use (10g in 1L of water)
  • USDA authorized for use in federally inspected meat and poultry plants

Used to clean:
Healthcare instruments, laboratory ware, vacuum equipment, tissue culture ware, personal protective equipment, sampling apparatus, catheters, tubing, pipes, radioactive contaminated articles, optical parts, electronic components, pharmaceutical apparatus, cosmetics manufacturing equipment, metal castings, forgings and stampings, industrial parts, tanks and reactors, passes inhibitory residue test for water analysis.

Used to remove:
Soil, grit, grime, buffing compound, slime, grease, oils, blood, tissue, salts, deposits, particulates, solvents, chemicals, radioisotopes, radioactive contaminations, silicon oils, mold release agents.

Surfaces cleaned:
Corrosion inhibited formulation recommended for glass, metal,stainless steel, porcelain, ceramic, plastic, rubber and fiberglass, can be used on soft metals such as copper, aluminum, zinc and magnesium if rinsed promptly.

Cleaning method:
Soak, brush, sponge, cloth, ultrasonic, flow through clean-inplace. Will foam—not for spray or machine use.

Shelf life 2 years from date of manufacture.

Order Info

0.5kg (1 lb.) makes 49.2L (13 gal.) cleaning solution.

Specifications

Product Type Powdered Precision Cleaner
Form Powder
pH 9.5
Appearance White, Cream
Container Type Carton
Content And Storage Store closed and upright in a cool dry place at 15° to 30°C.
For Use With (Application) Cleaning contaminants from glassware, metals, plastics, ceramic, porcelain, rubber and fiberglass
Volume (English) 50 lb.
Volume (Metric) 22.6 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.