Learn More
DWK Life Sciences Wheaton™ Power Accessories for Socorex™ Acura Electro Electronic Micropipettes
SDP
Catalog No. p-3070456 Shop All DWK Life Sciences Products
Change view
Click to view available options
Type:
Charging Cord
Charging Stand
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
13-709-78 Charging Cord
13-709-75 Charging Stand
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-709-78 Supplier DWK Life Sciences Supplier No. W820022A
Only null left
Add to Cart
Add to Cart
Stand allows charging and storing of up to three electronic pipettes or battery packs simultaneously.
  • Charging Stand for Pipettes or Batteries with 3 Positions
TRUSTED_SUSTAINABILITY
Specifications
Type Charging Cord
For Use With (Equipment) Socorex™ Acura™ Electro™ electronic micropipette
Voltage 110 V