Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Power Adapter, 24 V

Catalog No. 07203950 Shop All Corning Products
Change view
Click to view available options
For Use With (Equipment):
Corning™ Mini Microcentrifuge 6770
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
07-203-950 Corning™ Mini Microcentrifuge 6770
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-203-950 Supplier Corning™ Supplier No. 401025
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Corning™ Power Adapter, 24V for use with Corning Mini Microcentrifuge 6770

This is an 24V adapter for Corning™ LSE™ Mini Microcentrifuge (Cat. No. 6770).

TRUSTED_SUSTAINABILITY

Specifications

Type Power Adapter
For Use With (Equipment) Corning™ Mini Microcentrifuge 6770
No. per Pack 1/Pk.
Voltage 24 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.