Learn More
Waters Corp Power Cord, IEC C13, 10 A, 125 V, Taiwan
Catalog No. 502391726 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This power cord is designed for use with Waters laboratory equipment, providing reliable power connection. It features an IEC C13 connector and is rated for 10 A and 125 V, suitable for use in Taiwan.
  • Equipped with an IEC C13 connector.
  • Rated for 10 A current.
  • Designed for 125 V power systems.
  • 2.5 m in length.
  • Compatible with power outlets in Taiwan.
  • Suitable for various LC, MS, and data products.
Catalog No. 50-239-1726 Supplier Waters Corp Supplier No. 442000188
This power cord is designed for use with Waters laboratory equipment, providing reliable power connection. It features an IEC C13 connector and is rated for 10 A and 125 V, suitable for use in Taiwan.
  • Equipped with an IEC C13 connector.
  • Rated for 10 A current.
  • Designed for 125 V power systems.
  • 2.5 m in length.
  • Compatible with power outlets in Taiwan.
  • Suitable for various LC, MS, and data products.