Learn More
Hach Company Power cord kit for SC200
Catalog No. 5000207809 Shop All Hach Company Products
Encompass
Power cord kit (North American) for SC200The North American power cord kit for the SC200 contains a 6 foot power cord along with strain relief and additional connectors to connect your controller to standard 120V power
Catalog No. 50-002-07809 Supplier Hach Company Supplier No. 9202900
Power cord kit (North American) for SC200The North American power cord kit for the SC200 contains a 6 foot power cord along with strain relief and additional connectors to connect your controller to standard 120V power