Learn More
Leica Biosystems Power Cord NEMA 5-20 to IEC C19 for Leica instruments
Catalog No. NC1543070
Encompass_Preferred
Power cord for Leica instruments with a NEMA 5-20 (USA/Japan) plug and an IEC 60320 C19 connector. Supplied as an accessory or replacement cable to connect compatible instruments to a NEMA 5-20 mains outlet.
  • NEMA 5-20 plug for 125 V, 20 A mains outlets.
  • IEC 60320 C19 connector for compatible instrument inlets.
  • Suitable as an accessory or replacement power cable for laboratory instruments.
  • Designed for mains power delivery to higher-current equipment.
Catalog No. NC1543070 Supplier Leica Biosystems Supplier No. 14041149613
Power cord for Leica instruments with a NEMA 5-20 (USA/Japan) plug and an IEC 60320 C19 connector. Supplied as an accessory or replacement cable to connect compatible instruments to a NEMA 5-20 mains outlet.
  • NEMA 5-20 plug for 125 V, 20 A mains outlets.
  • IEC 60320 C19 connector for compatible instrument inlets.
  • Suitable as an accessory or replacement power cable for laboratory instruments.
  • Designed for mains power delivery to higher-current equipment.