Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gilson Power Cord US - JP

Catalog No. FH07011G Shop All Gilson Products
Change view
Click to view available options
Weight (English):
0.22 lb.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (English)
FH07011G 0.22 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FH07011G Supplier Gilson Supplier No. FH07011
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used to connect a Gilson instrument to mains power when the instrument requires a standard US/Japanese plug configuration

  • This is relevant for labs, institutions, or users who import Gilson equipment from US/Japan or who wish to maintain standardization using original manufacturer power cords.
  • Gilson instrument (lab equipment, pipette controller, etc.) to mains power when the instrument requires a standard US/Japanese plug configuration (i.e. 2-prong or 3-prong Type A/B plug, typically 100-125 VAC, 50–60 Hz) rather than a European, Indian, or other regional plug.

Specifications

Product Type Power Cord
Weight (English) 0.22 lb.
Weight (Metric) 0.1 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.