Learn More
Grainger Power Correction Capacitor 0.5 kvar
Catalog No. 502767818
Encompass_Preferred
The Power Correction Capacitor 0.5 kvar 16Y122 is a single-phase capacitor designed for power factor correction. It has a voltage rating of 240VAC and a capacitance of 0.5 kvar. This oil-filled capacitor features a self-healing design and is enclosed in a round steel case with stud terminals. It can help improve power factor and reduce energy costs in commercial and industrial applications when properly installed. This capacitor measures 5-1/4" diameter x 10-1/2" height and weighs approximately 9 lbs. It is UL and CSA listed.
Catalog No. 50-276-7818 Supplier Grainger Supplier No. 16Y122
The Power Correction Capacitor 0.5 kvar 16Y122 is a single-phase capacitor designed for power factor correction. It has a voltage rating of 240VAC and a capacitance of 0.5 kvar. This oil-filled capacitor features a self-healing design and is enclosed in a round steel case with stud terminals. It can help improve power factor and reduce energy costs in commercial and industrial applications when properly installed. This capacitor measures 5-1/4" diameter x 10-1/2" height and weighs approximately 9 lbs. It is UL and CSA listed.