Learn More
Grainger Power Correction Capacitor 0.75 kvar
Catalog No. 502766222
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a voltage rating of 240VAC and a capacitance value of 30 microfarads. It features a heavy duty canister construction and is UL and CSA listed. The capacitor helps reduce energy costs and prevent overheating and premature motor failure. It measures 5.5 inches by 5.5 inches and comes with mounting studs for secure installation. This capacitor is ideal for use with motors, transformers, ballasts, and other inductive loads up to 7.5 horsepower.
Catalog No. 50-276-6222 Supplier Grainger Supplier No. 16Y163
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a voltage rating of 240VAC and a capacitance value of 30 microfarads. It features a heavy duty canister construction and is UL and CSA listed. The capacitor helps reduce energy costs and prevent overheating and premature motor failure. It measures 5.5 inches by 5.5 inches and comes with mounting studs for secure installation. This capacitor is ideal for use with motors, transformers, ballasts, and other inductive loads up to 7.5 horsepower.