Learn More
Grainger Power Correction Capacitor 0.75 kvar
Catalog No. 502767163
Encompass_Preferred
The WEG Power Factor Correction Capacitor 16Y143 is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a capacitance of 143 microfarads and is rated for 480 volts AC. It features a heavy duty canister construction and is UL and CSA listed. The capacitor helps reduce energy costs and prevent overheating and inefficiency in motors, transformers, and inductive loads. This model is ideal for use in commercial and industrial applications.
Catalog No. 50-276-7163 Supplier Grainger Supplier No. 16Y143
The WEG Power Factor Correction Capacitor 16Y143 is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a capacitance of 143 microfarads and is rated for 480 volts AC. It features a heavy duty canister construction and is UL and CSA listed. The capacitor helps reduce energy costs and prevent overheating and inefficiency in motors, transformers, and inductive loads. This model is ideal for use in commercial and industrial applications.