Learn More
Grainger Power Correction Capacitor 1.5 kvar
Catalog No. 502765502
Encompass_Preferred
The WEG Power Factor Correction Capacitor 16Y125 is a single-phase capacitor designed to improve power factor in AC power systems. It has a capacitance of 125 microfarads with a voltage rating of 250 VAC. The capacitor uses metallized polypropylene film dielectric and is enclosed in a steel can. It can provide automatic power factor correction when used with appropriate controls. The capacitor measures 5.9" diameter x 7.9" height and weighs 9 lbs. It operates between -30°F to 165°F ambient temperature. The 16Y125 helps reduce energy costs and improves voltage regulation in motors, transformers, inductive loads. It's CSA certified and backed by a 12 month warranty from WEG.
Catalog No. 50-276-5502 Supplier Grainger Supplier No. 16Y125
The WEG Power Factor Correction Capacitor 16Y125 is a single-phase capacitor designed to improve power factor in AC power systems. It has a capacitance of 125 microfarads with a voltage rating of 250 VAC. The capacitor uses metallized polypropylene film dielectric and is enclosed in a steel can. It can provide automatic power factor correction when used with appropriate controls. The capacitor measures 5.9" diameter x 7.9" height and weighs 9 lbs. It operates between -30°F to 165°F ambient temperature. The 16Y125 helps reduce energy costs and improves voltage regulation in motors, transformers, inductive loads. It's CSA certified and backed by a 12 month warranty from WEG.