Learn More
Grainger Power Correction Capacitor 1.5 kvar
Catalog No. 502767846
Encompass_Preferred
The Grainger 16Y189 is a Westward 3/8-inch drive impact socket adapter that converts a 3/8-inch drive to a 1/2-inch drive. It is made from chrome vanadium steel for durability and has a black phosphate finish to resist corrosion. This impact socket adapter measures 2-1/2 inches long and weighs 0.19 lbs. It is rated for up to 10,000 ft-lbs of torque. The Westward 16Y189 socket adapter allows use of 3/8-inch drive tools with 1/2-inch drive impact wrenches.
Catalog No. 50-276-7846 Supplier Grainger Supplier No. 16Y189
The Grainger 16Y189 is a Westward 3/8-inch drive impact socket adapter that converts a 3/8-inch drive to a 1/2-inch drive. It is made from chrome vanadium steel for durability and has a black phosphate finish to resist corrosion. This impact socket adapter measures 2-1/2 inches long and weighs 0.19 lbs. It is rated for up to 10,000 ft-lbs of torque. The Westward 16Y189 socket adapter allows use of 3/8-inch drive tools with 1/2-inch drive impact wrenches.