Learn More
Grainger Power Correction Capacitor 10 kvar
Catalog No. 502765059
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a kVAR rating of 7.5 with a voltage rating of 240VAC. It features an internal discharge resistor for safety and has UL and CSA certifications. The capacitor uses dry polypropylene film dielectric material in a sealed steel enclosure for durability. It measures 10.25" H x 5.75" W x 3.5" D and weighs 7 lbs. This capacitor helps reduce energy waste and improves voltage regulation in motors, transformers, lighting, and other inductive loads. It's ideal for commercial and industrial applications.
Catalog No. 50-276-5059 Supplier Grainger Supplier No. 16Y195
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a kVAR rating of 7.5 with a voltage rating of 240VAC. It features an internal discharge resistor for safety and has UL and CSA certifications. The capacitor uses dry polypropylene film dielectric material in a sealed steel enclosure for durability. It measures 10.25" H x 5.75" W x 3.5" D and weighs 7 lbs. This capacitor helps reduce energy waste and improves voltage regulation in motors, transformers, lighting, and other inductive loads. It's ideal for commercial and industrial applications.