Learn More
Grainger Power Correction Capacitor 12.5 kvar
Catalog No. 502767795
Encompass_Preferred
The 16Y152 power factor correction capacitor from Grainger is a single-phase capacitor rated for 480 volts with a 150 microfarad capacitance. It is designed for use in AC power systems to improve power factor and reduce energy costs. This cylindrical run capacitor has Quick Connect terminals and is UL and CSA listed. It can help reduce peak demand charges and correct power factor in motors, transformers, inductive lighting, and other inductive loads. With dimensions of 4-1/2" diameter x 10-1/2" height and a weight of 9 lbs, this capacitor provides economical power factor correction.
Catalog No. 50-276-7795 Supplier Grainger Supplier No. 16Y152
The 16Y152 power factor correction capacitor from Grainger is a single-phase capacitor rated for 480 volts with a 150 microfarad capacitance. It is designed for use in AC power systems to improve power factor and reduce energy costs. This cylindrical run capacitor has Quick Connect terminals and is UL and CSA listed. It can help reduce peak demand charges and correct power factor in motors, transformers, inductive lighting, and other inductive loads. With dimensions of 4-1/2" diameter x 10-1/2" height and a weight of 9 lbs, this capacitor provides economical power factor correction.