Learn More
Grainger Power Correction Capacitor 12.5 kvar
Catalog No. 502767799
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. It has a 480VAC voltage rating and is available in sizes from 7.5kVAR to 450kVAR. The capacitor uses dry polypropylene film dielectric material and is enclosed in a vented steel enclosure. It can provide automatic or manual power factor correction. This capacitor helps reduce energy consumption and improves voltage regulation. Its applications include industrial facilities, commercial buildings, utilities, and renewable energy systems. The capacitor offers reliable performance, safety, and long service life.
Catalog No. 50-276-7799 Supplier Grainger Supplier No. 16Y196
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. It has a 480VAC voltage rating and is available in sizes from 7.5kVAR to 450kVAR. The capacitor uses dry polypropylene film dielectric material and is enclosed in a vented steel enclosure. It can provide automatic or manual power factor correction. This capacitor helps reduce energy consumption and improves voltage regulation. Its applications include industrial facilities, commercial buildings, utilities, and renewable energy systems. The capacitor offers reliable performance, safety, and long service life.