Learn More
Grainger Power Correction Capacitor 20 kvar
Catalog No. 502766215
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. It has a 480VAC voltage rating and is available in sizes from 7.5kVAR to 450kVAR. The capacitor uses dry polypropylene film dielectric material and is enclosed in a vented steel enclosure. It can provide power factor correction for motors, transformers, lighting, and other inductive loads. The capacitor should be installed with external fuses or breakers for overload protection. Regular maintenance includes inspection, cleaning, and testing.
Catalog No. 50-276-6215 Supplier Grainger Supplier No. 16Y135
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. It has a 480VAC voltage rating and is available in sizes from 7.5kVAR to 450kVAR. The capacitor uses dry polypropylene film dielectric material and is enclosed in a vented steel enclosure. It can provide power factor correction for motors, transformers, lighting, and other inductive loads. The capacitor should be installed with external fuses or breakers for overload protection. Regular maintenance includes inspection, cleaning, and testing.