Learn More
Grainger Power Correction Capacitor 20 kvar
Catalog No. 502766226
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a voltage rating of 240VAC and a capacitance value of 30 microfarads. It features a heavy duty canister construction and is UL and CSA certified. The capacitor helps reduce energy costs and prevent overheating and premature motor failure. It measures 5.9" diameter x 10.6" height and weighs 9 lbs. This 30 microfarad capacitor is well-suited for use in commercial and industrial applications to improve power factor to 0.95 or better.
Catalog No. 50-276-6226 Supplier Grainger Supplier No. 16Y155
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a voltage rating of 240VAC and a capacitance value of 30 microfarads. It features a heavy duty canister construction and is UL and CSA certified. The capacitor helps reduce energy costs and prevent overheating and premature motor failure. It measures 5.9" diameter x 10.6" height and weighs 9 lbs. This 30 microfarad capacitor is well-suited for use in commercial and industrial applications to improve power factor to 0.95 or better.