Learn More
Grainger Power Correction Capacitor 20 kvar
Catalog No. 502766984
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a kVAR rating of 25 kVAR with 480V voltage. It features an internal discharge resistor for safety and has an epoxy-filled aluminum can enclosure. The dimensions are 14" diameter x 24" height. This capacitor helps reduce energy waste and improves voltage regulation when installed on the load side of motors, transformers, induction furnaces, and other inductive equipment. It is UL and CSA listed.
Catalog No. 50-276-6984 Supplier Grainger Supplier No. 16Y175
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a kVAR rating of 25 kVAR with 480V voltage. It features an internal discharge resistor for safety and has an epoxy-filled aluminum can enclosure. The dimensions are 14" diameter x 24" height. This capacitor helps reduce energy waste and improves voltage regulation when installed on the load side of motors, transformers, induction furnaces, and other inductive equipment. It is UL and CSA listed.