Learn More
Grainger Power Correction Capacitor 22.5 kvar
Catalog No. 502766827
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. It has a 480VAC voltage rating and is available in sizes from 7.5kVAR to 450kVAR. The capacitor uses dry polypropylene film dielectric material and is enclosed in a vented steel enclosure for safety. It can provide leading vars to offset inductive loads and improve power factor to 0.95 or better. This helps reduce utility costs and improves voltage regulation. It is CSA certified and backed by a 12 month warranty from WEG. The capacitor is designed for indoor installations and has built-in discharge resistors for safety during servicing.
Catalog No. 50-276-6827 Supplier Grainger Supplier No. 16Y201
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. It has a 480VAC voltage rating and is available in sizes from 7.5kVAR to 450kVAR. The capacitor uses dry polypropylene film dielectric material and is enclosed in a vented steel enclosure for safety. It can provide leading vars to offset inductive loads and improve power factor to 0.95 or better. This helps reduce utility costs and improves voltage regulation. It is CSA certified and backed by a 12 month warranty from WEG. The capacitor is designed for indoor installations and has built-in discharge resistors for safety during servicing.