Learn More
Grainger Power Correction Capacitor 25 kvar
Catalog No. 502765546
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a capacitance of 25 microfarads with a voltage rating of 440VAC. It features a heavy duty canister construction and is UL and CSA certified. The dimensions are 7" diameter x 17" height and it weighs approximately 35 lbs. This capacitor helps reduce energy costs and prevent overloading by correcting power factor issues. It is well-suited for commercial and industrial applications.
Catalog No. 50-276-5546 Supplier Grainger Supplier No. 16Y136
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a capacitance of 25 microfarads with a voltage rating of 440VAC. It features a heavy duty canister construction and is UL and CSA certified. The dimensions are 7" diameter x 17" height and it weighs approximately 35 lbs. This capacitor helps reduce energy costs and prevent overloading by correcting power factor issues. It is well-suited for commercial and industrial applications.