Learn More
Grainger Power Correction Capacitor 25 kvar
Catalog No. 502767165
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a 3-phase capacitor designed to improve power factor and reduce energy costs. It has a 480VAC voltage rating, 200kVAR power rating, and 60Hz frequency. This heavy duty capacitor features a steel enclosure and is UL and CSA certified. It can help reduce peak demand charges and correct power factor to 0.95 or better. This capacitor would be well-suited for large industrial and commercial applications.
Catalog No. 50-276-7165 Supplier Grainger Supplier No. 16Y156
The WEG Power Factor Correction Capacitor is a 3-phase capacitor designed to improve power factor and reduce energy costs. It has a 480VAC voltage rating, 200kVAR power rating, and 60Hz frequency. This heavy duty capacitor features a steel enclosure and is UL and CSA certified. It can help reduce peak demand charges and correct power factor to 0.95 or better. This capacitor would be well-suited for large industrial and commercial applications.