Learn More
Grainger Power Correction Capacitor 27.5 kvar
Catalog No. 502765075
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a 3-phase capacitor designed to improve power factor and reduce energy costs. This capacitor has a voltage rating of 480VAC and a capacitance of 150 microfarads. It features a heavy duty steel enclosure and is UL and CSA certified. The capacitor helps reduce kVAR demand and improve power factor to 0.95 or better. It is suitable for use in industrial and commercial applications.
Catalog No. 50-276-5075 Supplier Grainger Supplier No. 16Y137
The WEG Power Factor Correction Capacitor is a 3-phase capacitor designed to improve power factor and reduce energy costs. This capacitor has a voltage rating of 480VAC and a capacitance of 150 microfarads. It features a heavy duty steel enclosure and is UL and CSA certified. The capacitor helps reduce kVAR demand and improve power factor to 0.95 or better. It is suitable for use in industrial and commercial applications.