Learn More
Grainger Power Correction Capacitor 27.5 kvar
Supplier:  Grainger 16Y203
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. It has a 480VAC voltage rating and is available in sizes from 7.5kVAR to 450kVAR. The capacitor uses dry polypropylene film dielectric material and is enclosed in a vented steel enclosure for safety. It can provide leading vars to offset inductive loads and improve power factor to 0.95 or better. This helps reduce utility costs and improves voltage regulation. It is CSA certified and backed by a 12 month warranty from WEG. The capacitor is designed for indoor installations and has built-in discharge resistors for safety during servicing.
Catalog No. 50-276-6184
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.