Learn More
Grainger Power Correction Capacitor 30 kvar
Catalog No. 502767005
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a kVAR rating of 7.5 with a voltage rating of 240VAC. It features an internal discharge resistor for safety and has UL and CSA certifications. The capacitor uses dry polypropylene film dielectric and is enclosed in a vented steel case. It can help reduce utility costs and improve voltage conditions when properly applied. This capacitor would be a good choice for power factor correction in small commercial or industrial applications.
Catalog No. 50-276-7005 Supplier Grainger Supplier No. 16Y179
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a kVAR rating of 7.5 with a voltage rating of 240VAC. It features an internal discharge resistor for safety and has UL and CSA certifications. The capacitor uses dry polypropylene film dielectric and is enclosed in a vented steel case. It can help reduce utility costs and improve voltage conditions when properly applied. This capacitor would be a good choice for power factor correction in small commercial or industrial applications.