Learn More
Grainger Power Correction Capacitor 35 kvar
Catalog No. 502766185
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a capacitance of 25 microfarads with a voltage rating of 440VAC. It features a heavy duty canister construction and is UL and CSA certified. The dimensions are 7" diameter x 17" height and it weighs approximately 35 lbs. This capacitor helps reduce energy waste and prevent overloading of transformers. It is well suited for use in commercial and industrial applications.
Catalog No. 50-276-6185 Supplier Grainger Supplier No. 16Y159
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a capacitance of 25 microfarads with a voltage rating of 440VAC. It features a heavy duty canister construction and is UL and CSA certified. The dimensions are 7" diameter x 17" height and it weighs approximately 35 lbs. This capacitor helps reduce energy waste and prevent overloading of transformers. It is well suited for use in commercial and industrial applications.