Learn More
Grainger Power Correction Capacitor 35 kvar
Catalog No. 502766830
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. It has a voltage rating of 240VAC and a capacitance of 30 microfarads. This oil-filled run capacitor features a rectangular steel case and is UL and CSA certified. It can help reduce energy costs and prevent overloading by correcting power factor issues in motors, transformers, inductive loads. With dimensions of 4-1/4" x 2-1/2" x 3-3/4", this capacitor provides economical power factor correction for residential and light commercial applications.
Catalog No. 50-276-6830 Supplier Grainger Supplier No. 16Y205
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. It has a voltage rating of 240VAC and a capacitance of 30 microfarads. This oil-filled run capacitor features a rectangular steel case and is UL and CSA certified. It can help reduce energy costs and prevent overloading by correcting power factor issues in motors, transformers, inductive loads. With dimensions of 4-1/4" x 2-1/2" x 3-3/4", this capacitor provides economical power factor correction for residential and light commercial applications.