Learn More
Grainger Power Correction Capacitor 40 kvar
Catalog No. 502767162
Encompass_Preferred
The WEG Power Factor Correction Capacitor 16Y140 is a single-phase power factor correction capacitor designed for 208-240V applications. It has a kVAR rating of 140 with a voltage rating of 240V. The capacitor helps improve power factor to 0.95 lead and uses dry polypropylene film dielectric. It has an operating temperature range of -40°F to 185°F and includes pressure-sensitive interrupters. The dimensions are 18"H x 14"W x 8.5"D. This capacitor aims to reduce energy consumption and improve power quality in industrial applications.
Catalog No. 50-276-7162 Supplier Grainger Supplier No. 16Y140
The WEG Power Factor Correction Capacitor 16Y140 is a single-phase power factor correction capacitor designed for 208-240V applications. It has a kVAR rating of 140 with a voltage rating of 240V. The capacitor helps improve power factor to 0.95 lead and uses dry polypropylene film dielectric. It has an operating temperature range of -40°F to 185°F and includes pressure-sensitive interrupters. The dimensions are 18"H x 14"W x 8.5"D. This capacitor aims to reduce energy consumption and improve power quality in industrial applications.