Learn More
Grainger Power Correction Capacitor 45 kvar
Catalog No. 502766987
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a 3-phase capacitor designed to improve power factor and reduce energy costs. This capacitor has a voltage rating of 480VAC and a capacitance of 150 microfarads. It features a heavy duty steel enclosure and is UL and CSA listed. The dimensions are 18"W x 24"H x 8"D. This capacitor can help reduce peak demand charges and improve voltage conditions in industrial and commercial applications. It requires a potential relay for protection. Overall, this is a heavy-duty power factor correction capacitor well-suited for larger industrial loads.
Catalog No. 50-276-6987 Supplier Grainger Supplier No. 16Y207
The WEG Power Factor Correction Capacitor is a 3-phase capacitor designed to improve power factor and reduce energy costs. This capacitor has a voltage rating of 480VAC and a capacitance of 150 microfarads. It features a heavy duty steel enclosure and is UL and CSA listed. The dimensions are 18"W x 24"H x 8"D. This capacitor can help reduce peak demand charges and improve voltage conditions in industrial and commercial applications. It requires a potential relay for protection. Overall, this is a heavy-duty power factor correction capacitor well-suited for larger industrial loads.