Learn More
Grainger Power Correction Capacitor 45 kvar
Catalog No. 502766978
Encompass_Preferred
The WEG Power Factor Correction Capacitor 16Y161 is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a capacitance of 161 microfarads and is rated for 480 volts AC. It features a heavy duty canister construction and is UL and CSA listed. The capacitor helps reduce energy costs and prevent overheating and inefficiency in motors, transformers, and inductive loads. This model is ideal for use in commercial and industrial applications.
Catalog No. 50-276-6978 Supplier Grainger Supplier No. 16Y161
The WEG Power Factor Correction Capacitor 16Y161 is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a capacitance of 161 microfarads and is rated for 480 volts AC. It features a heavy duty canister construction and is UL and CSA listed. The capacitor helps reduce energy costs and prevent overheating and inefficiency in motors, transformers, and inductive loads. This model is ideal for use in commercial and industrial applications.