Learn More
Grainger Power Correction Capacitor 45 kvar
Catalog No. 502767798
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a kVAR rating of 7.5 with a voltage rating of 240VAC. It features an internal discharge resistor for safety and has an operating temperature range of -40°F to 140°F. The capacitor uses dry polypropylene film dielectric material and comes in a NEMA 1 steel enclosure suitable for indoor installations. It can help reduce energy consumption and utility costs when properly applied. The dimensions are 10" H x 8" W x 8" D and it weighs 13 lbs.
Catalog No. 50-276-7798 Supplier Grainger Supplier No. 16Y141
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a kVAR rating of 7.5 with a voltage rating of 240VAC. It features an internal discharge resistor for safety and has an operating temperature range of -40°F to 140°F. The capacitor uses dry polypropylene film dielectric material and comes in a NEMA 1 steel enclosure suitable for indoor installations. It can help reduce energy consumption and utility costs when properly applied. The dimensions are 10" H x 8" W x 8" D and it weighs 13 lbs.