Learn More
Grainger Power Correction Capacitor 60 kvar
Catalog No. 502767794
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a kVAR rating of 25 kVAR with 480 VAC voltage. It features an internal discharge resistor for safety and has an epoxy-filled aluminum can enclosure. The operating temperature range is -40°F to 140°F. This capacitor measures 14 in x 14 in x 12 in and weighs approximately 65 lbs. It can help reduce energy consumption and improve voltage conditions when properly installed in parallel with inductive loads.
Catalog No. 50-276-7794 Supplier Grainger Supplier No. 16Y184
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a kVAR rating of 25 kVAR with 480 VAC voltage. It features an internal discharge resistor for safety and has an epoxy-filled aluminum can enclosure. The operating temperature range is -40°F to 140°F. This capacitor measures 14 in x 14 in x 12 in and weighs approximately 65 lbs. It can help reduce energy consumption and improve voltage conditions when properly installed in parallel with inductive loads.