Learn More
Grainger Power Correction Capacitor 7.5 kvar
Catalog No. 502767848
Encompass_Preferred
The WEG Power Factor Correction Capacitor 16Y170 is a single-phase capacitor designed to improve power factor in electrical systems. It has a capacitance of 170 microfarads and is rated for 480 volts AC. The capacitor uses metallized polypropylene film dielectric and is enclosed in a steel can. It can provide automatic power factor correction when used with appropriate controls. The capacitor has UL and CSA certifications and is manufactured in Brazil by WEG, a global manufacturer of electric motors, drives, and controls. With its high capacitance and voltage rating, this capacitor can efficiently correct power factor issues in many commercial and industrial applications.
Catalog No. 50-276-7848 Supplier Grainger Supplier No. 16Y170
The WEG Power Factor Correction Capacitor 16Y170 is a single-phase capacitor designed to improve power factor in electrical systems. It has a capacitance of 170 microfarads and is rated for 480 volts AC. The capacitor uses metallized polypropylene film dielectric and is enclosed in a steel can. It can provide automatic power factor correction when used with appropriate controls. The capacitor has UL and CSA certifications and is manufactured in Brazil by WEG, a global manufacturer of electric motors, drives, and controls. With its high capacitance and voltage rating, this capacitor can efficiently correct power factor issues in many commercial and industrial applications.