Learn More
Grainger Power Correction Capacitor 75 kvar
Catalog No. 502767160
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a kVAR rating of 25 kVAR with 480V voltage. It features a heavy duty canister construction and is UL listed. The dimensions are 18" diameter x 39" height and it weighs 285 lbs. This 3-phase power factor correction capacitor helps reduce energy costs and improves voltage conditions in facilities with inductive loads. It is suitable for industrial and commercial applications.
Catalog No. 50-276-7160 Supplier Grainger Supplier No. 16Y210
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a kVAR rating of 25 kVAR with 480V voltage. It features a heavy duty canister construction and is UL listed. The dimensions are 18" diameter x 39" height and it weighs 285 lbs. This 3-phase power factor correction capacitor helps reduce energy costs and improves voltage conditions in facilities with inductive loads. It is suitable for industrial and commercial applications.