Learn More
MSC TPI Corporation Disconnect Switch, 3-Pole, 100 A, 120 V to 600 V, 1 ea
Catalog No. 5000507342
Encompass
This power disconnect switch is designed for use with 5100 series horizontal or vertical mounted fan forced unit heaters. It provides a reliable method for isolating power to the heating unit during maintenance or repair.
  • Rated for 100 A current
  • Configured with 3 poles
  • Supports a voltage range of 120 VAC to 600 VAC
  • Compatible with 5100 series fan forced unit heaters
  • Full rated interrupt capacity
Catalog No. 50-005-07342 Supplier MSC Supplier No. 77989960
This power disconnect switch is designed for use with 5100 series horizontal or vertical mounted fan forced unit heaters. It provides a reliable method for isolating power to the heating unit during maintenance or repair.
  • Rated for 100 A current
  • Configured with 3 poles
  • Supports a voltage range of 120 VAC to 600 VAC
  • Compatible with 5100 series fan forced unit heaters
  • Full rated interrupt capacity