Learn More
MSC Tpi Corporation Power Disconnect Switch, 3-Pole, 40 Amp, 5100 Series, 1 ea
Catalog No. 5000507343
Encompass
This 3-pole, 40 A power disconnect switch is intended for use with 5100 series horizontal or vertical mounted fan-forced unit heaters. It provides a means to disconnect power for maintenance or emergency shutdown in indoor industrial and commercial environments.
  • Designed for use with 5100 series fan-forced unit heaters
  • Rated for 40 A
  • Three-pole configuration
  • Supports voltages from 120 to 600 VAC
  • Manufactured in the U.S.A.
Catalog No. 50-005-07343 Supplier MSC Supplier No. 77989986
This 3-pole, 40 A power disconnect switch is intended for use with 5100 series horizontal or vertical mounted fan-forced unit heaters. It provides a means to disconnect power for maintenance or emergency shutdown in indoor industrial and commercial environments.
  • Designed for use with 5100 series fan-forced unit heaters
  • Rated for 40 A
  • Three-pole configuration
  • Supports voltages from 120 to 600 VAC
  • Manufactured in the U.S.A.