Learn More
MSC Tpi Power Disconnect Switch, 2-Pole, 20 A, 5100 Series, 1 ea
Catalog No. 5000507388
Encompass
This non-fused pull-out AC disconnect switch is designed for use with 5100 series horizontal or vertical mounted fan-forced unit heaters. It provides a reliable means to disconnect power for maintenance or safety purposes in commercial and industrial settings.
  • Two-pole configuration
  • Compatible with 5100 series unit heaters
  • Rated for 120 V to 277 VAC
  • Interrupt rating of 10,000 A
  • UL/CUL listed for safety
Catalog No. 50-005-07388 Supplier MSC Supplier No. 77989978
This non-fused pull-out AC disconnect switch is designed for use with 5100 series horizontal or vertical mounted fan-forced unit heaters. It provides a reliable means to disconnect power for maintenance or safety purposes in commercial and industrial settings.
  • Two-pole configuration
  • Compatible with 5100 series unit heaters
  • Rated for 120 V to 277 VAC
  • Interrupt rating of 10,000 A
  • UL/CUL listed for safety