Learn More
Grainger Power Factor Correction Capacitor 5 kvar
Catalog No. 502767847
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a voltage rating of 240VAC and a capacitance value of 30 microfarads. It features a heavy duty canister construction and is UL and CSA listed. The dimensions are 7" diameter x 17" height and it weighs approximately 35 lbs. This capacitor helps reduce energy consumption and improve voltage conditions in motors, transformers, lighting and other inductive loads. It would be well-suited for commercial and industrial applications requiring power factor correction.
Catalog No. 50-276-7847 Supplier Grainger Supplier No. 16Y169
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a voltage rating of 240VAC and a capacitance value of 30 microfarads. It features a heavy duty canister construction and is UL and CSA listed. The dimensions are 7" diameter x 17" height and it weighs approximately 35 lbs. This capacitor helps reduce energy consumption and improve voltage conditions in motors, transformers, lighting and other inductive loads. It would be well-suited for commercial and industrial applications requiring power factor correction.