Learn More
Grainger Power Factor Correction Capacitor 3 kvar
Catalog No. 502766832
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a voltage rating of 240VAC and a capacitance value of 30 microfarads. It features a heavy duty canister construction and is UL and CSA listed. The capacitor helps reduce energy costs and prevent overheating and premature motor failure. This 30 microfarad capacitor is well suited for use with 1 to 3 HP motors.
Catalog No. 50-276-6832 Supplier Grainger Supplier No. 16Y192
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a voltage rating of 240VAC and a capacitance value of 30 microfarads. It features a heavy duty canister construction and is UL and CSA listed. The capacitor helps reduce energy costs and prevent overheating and premature motor failure. This 30 microfarad capacitor is well suited for use with 1 to 3 HP motors.