Learn More
Grainger Power Factor Correction Capacitor 1 kvar
Catalog No. 502767007
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a 3-phase capacitor designed to improve power factor and reduce energy costs. It has a 480VAC voltage rating, 200kVAR power rating, 60Hz frequency, and features an internal discharge resistor with terminals. The capacitor uses dry, self-healing metallized polypropylene film dielectric and is UL listed. It comes in a NEMA 1 indoor steel enclosure suitable for wall, floor, or rack mounting. This capacitor can help reduce utility penalties and improve voltage conditions in industrial applications.
Catalog No. 50-276-7007 Supplier Grainger Supplier No. 16Y124
The WEG Power Factor Correction Capacitor is a 3-phase capacitor designed to improve power factor and reduce energy costs. It has a 480VAC voltage rating, 200kVAR power rating, 60Hz frequency, and features an internal discharge resistor with terminals. The capacitor uses dry, self-healing metallized polypropylene film dielectric and is UL listed. It comes in a NEMA 1 indoor steel enclosure suitable for wall, floor, or rack mounting. This capacitor can help reduce utility penalties and improve voltage conditions in industrial applications.