Learn More
Grainger Power Factor Correction Capacitor 2 kvar
Catalog No. 502767843
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. It has a voltage rating of 240VAC and a capacitance of 30 microfarads. The capacitor is enclosed in a steel can with epoxy resin end bells and uses metallized polypropylene film dielectric. It can operate at temperatures between -40°F to 185°F. This capacitor helps reduce energy costs and improve voltage regulation by correcting power factor issues. It is UL and CSA listed.
Catalog No. 50-276-7843 Supplier Grainger Supplier No. 16Y166
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. It has a voltage rating of 240VAC and a capacitance of 30 microfarads. The capacitor is enclosed in a steel can with epoxy resin end bells and uses metallized polypropylene film dielectric. It can operate at temperatures between -40°F to 185°F. This capacitor helps reduce energy costs and improve voltage regulation by correcting power factor issues. It is UL and CSA listed.