Learn More
Grainger Power Factor Correction Capacitor 1 kvar
Catalog No. 502766216
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. It has a voltage rating of 240VAC and a capacitance of 30 microfarads. The capacitor helps reduce energy waste and lower utility costs by correcting power factor issues. This model has an enclosure rating of NEMA 1 and includes pressure disconnecting devices for safety. It can provide automatic power factor correction when used with appropriate controls. The capacitor has UL and CSA certifications.
Catalog No. 50-276-6216 Supplier Grainger Supplier No. 16Y144
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. It has a voltage rating of 240VAC and a capacitance of 30 microfarads. The capacitor helps reduce energy waste and lower utility costs by correcting power factor issues. This model has an enclosure rating of NEMA 1 and includes pressure disconnecting devices for safety. It can provide automatic power factor correction when used with appropriate controls. The capacitor has UL and CSA certifications.