Learn More
Grainger Power Factor Correction Capacitor 1 kvar
Catalog No. 502766855
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. It has a voltage rating of 240VAC and a capacitance of 30 microfarads. The capacitor is enclosed in a steel enclosure with epoxy resin insulation and uses self-healing metallized film technology. It can handle temperatures between -25°C to 70°C and has quick-connect terminals for easy installation. This capacitor helps reduce energy costs and improves power quality in lighting, motor, and other inductive load applications. With its durable construction, it provides reliable power factor correction over time.
Catalog No. 50-276-6855 Supplier Grainger Supplier No. 16Y164
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. It has a voltage rating of 240VAC and a capacitance of 30 microfarads. The capacitor is enclosed in a steel enclosure with epoxy resin insulation and uses self-healing metallized film technology. It can handle temperatures between -25°C to 70°C and has quick-connect terminals for easy installation. This capacitor helps reduce energy costs and improves power quality in lighting, motor, and other inductive load applications. With its durable construction, it provides reliable power factor correction over time.