Learn More
Grainger Power Factor Correction Capacitor 5 kvar
Catalog No. 502766981
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a kVAR rating of 7.5 with a voltage rating of 240VAC. It features an internal discharge resistor for safety and has UL and CSA certifications. The capacitor uses dry polypropylene film dielectric material in a sealed steel enclosure for durability. It can help reduce utility costs and improve voltage conditions when properly installed in parallel with inductive loads. This capacitor measures 10.25 x 6 x 12.25 inches and weighs 13 pounds. It is well-suited for use in commercial and industrial applications.
Catalog No. 50-276-6981 Supplier Grainger Supplier No. 16Y193
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a kVAR rating of 7.5 with a voltage rating of 240VAC. It features an internal discharge resistor for safety and has UL and CSA certifications. The capacitor uses dry polypropylene film dielectric material in a sealed steel enclosure for durability. It can help reduce utility costs and improve voltage conditions when properly installed in parallel with inductive loads. This capacitor measures 10.25 x 6 x 12.25 inches and weighs 13 pounds. It is well-suited for use in commercial and industrial applications.