Learn More
Grainger Power Factor Correction Capacitor 3 kvar
Catalog No. 502765545
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a kVAR rating of 25 kVAR with 480 VAC voltage. It features a heavy-duty steel enclosure and is UL and CSA certified. The dimensions are 24" H x 14" W x 8" D. This capacitor helps reduce energy costs and prevent overloading by correcting power factor issues. It is well-suited for commercial and industrial applications.
Catalog No. 50-276-5545 Supplier Grainger Supplier No. 16Y168
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a kVAR rating of 25 kVAR with 480 VAC voltage. It features a heavy-duty steel enclosure and is UL and CSA certified. The dimensions are 24" H x 14" W x 8" D. This capacitor helps reduce energy costs and prevent overloading by correcting power factor issues. It is well-suited for commercial and industrial applications.