Learn More
Grainger Power Factor Correction Capacitor 3 kvar
Catalog No. 502767826
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a voltage rating of 240VAC and a capacitance value of 30 microfarads. It features a heavy duty canister construction and is UL and CSA listed. The dimensions are 7" diameter x 14" height and it weighs approximately 35 lbs. This capacitor helps reduce energy waste and prevent overloading of transformers. It's commonly used in commercial and industrial applications.
Catalog No. 50-276-7826 Supplier Grainger Supplier No. 16Y148
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a voltage rating of 240VAC and a capacitance value of 30 microfarads. It features a heavy duty canister construction and is UL and CSA listed. The dimensions are 7" diameter x 14" height and it weighs approximately 35 lbs. This capacitor helps reduce energy waste and prevent overloading of transformers. It's commonly used in commercial and industrial applications.