Learn More
Grainger Power Factor Correction Capacitor 2 kvar
Catalog No. 502767159
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a kVAR rating of 25 kVAR with 480 VAC voltage. It features an internal discharge resistor for safety and has an epoxy-filled aluminum can enclosure. The operating temperature range is -40°F to 140°F. This capacitor measures 14 in x 14 in x 12 in and weighs approximately 65 lbs. It can help reduce energy consumption and improve voltage conditions when properly installed in parallel with inductive loads. This product is UL and CSA listed.
Catalog No. 50-276-7159 Supplier Grainger Supplier No. 16Y126
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a kVAR rating of 25 kVAR with 480 VAC voltage. It features an internal discharge resistor for safety and has an epoxy-filled aluminum can enclosure. The operating temperature range is -40°F to 140°F. This capacitor measures 14 in x 14 in x 12 in and weighs approximately 65 lbs. It can help reduce energy consumption and improve voltage conditions when properly installed in parallel with inductive loads. This product is UL and CSA listed.