Learn More
Ace Glass, Inc. POWER HOLD CLAMP COMPLETE POWER HOLD CLAMP COMPLETE
SDP
Catalog No. 509558906
Encompass
Power hold clamp by Arrow. Clamp fits support stand with 3/8¿ ¿ 5/8¿ diameter shaft and stirrers with mounting rod from 3/8¿ ¿ 5/8¿ diameter. Includes stop collar.
Catalog No. 50-955-8906 Supplier Ace Glass, Inc. Supplier No. 1108207
Power hold clamp by Arrow. Clamp fits support stand with 3/8¿ ¿ 5/8¿ diameter shaft and stirrers with mounting rod from 3/8¿ ¿ 5/8¿ diameter. Includes stop collar.