Learn More
Arrow Engineering PHC-750 Power Hold Clamp
Catalog No. NC1420491
Encompass_Preferred
The Arrow Mixing Products PHC-750 fits support stands from 1/2″ to 3/4″ dia. (12 mm to 19 mm) and can be used with Stand ASE-750 for a quick, complete laboratory mixer setup. It is is engineered for precision and durability, making it an essential tool for securing various materials during mixing and processing applications.
Catalog No. NC1420491 Supplier Arrow Engineering Supplier No. PHC750
The Arrow Mixing Products PHC-750 fits support stands from 1/2″ to 3/4″ dia. (12 mm to 19 mm) and can be used with Stand ASE-750 for a quick, complete laboratory mixer setup. It is is engineered for precision and durability, making it an essential tool for securing various materials during mixing and processing applications.