Learn More
Brady™ Power Insertion Field Wiring I/O Adapter for Virtual Sign Projector
Shop All Brady Corporation ProductsDescription
- Powers the sensor and routes its control wire without requiring you to cut the projector's connector.
- Tool-free solution with convenient screw terminals for easy field installation.
- One male M15 to driver DIM+/- input and one female M15 to driver VDC output.
- Accepts any DC input voltage up to 24V.
- Input terminals: 1 = DIM+, 2 = DIM-, 3 = Power+, 4 = Power-.
- For use with compatible microwave motion and infrared presence sensors for virtual sign projectors.
Specifications
Specifications
| Product Type | Cable |
| For Use With (Equipment) | Virtual Sign Projector |
| Height (English) | 72 in. |
| Diameter (English) | 1 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.